Order Entry
Sweden
ContactUsLinkComponent
Human recombinant TGF beta 3 (from E. coli)
Human recombinant TGF beta 3 (from E. coli)
Catalog # PRSI40-574
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TGF beta 3 (from E. coli)
Catalog # PRSI40-574
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Biological activity was determined by TGF-β3’s ability to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells. The expected ED50 is ≤ 0.05 ng/mL, corresponding to a specific activity of ≥2 x 107 units/mg. Endotoxin level is less than 0.1 ng per μg of TGF-β3 (1EU/μg).
  • Gene ID:
    7043
  • Protein synonyms:
    Transforming growth factor beta-3|TGF-beta-3|RNHF|ARVD|ARVD1|TGF-beta3
  • Protein/peptide name:
    TGF beta 3
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications

About this item

The three mammalian isoforms of TGF-β, TGF-β1, β2, β3, signal through the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and would healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF-β isoform from a latent complex involves proteolytic processing of the complex and /or induction of conformational changes by proteins such as thrombospondin-1. The physiological role of TGF-β3 is still unknown but its expression pattern suggests a role in the regulation of certain development processes. Recombinant TGF-β3 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond.Manufactured using all non-animal reagents.

  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.