Order Entry
Sweden
ContactUsLinkComponent
Human recombinant IL7 (from E. coli)
Human recombinant IL7 (from E. coli)
Catalog # PRSI40-561
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL7 (from E. coli)
Catalog # PRSI40-561
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Biological activity was determined by its ability to stimulate the proliferation of mouse 2E8 cells. The expected ED50 is ≤0.5 ng/mL, corresponding to a specific activity of ≥2 x 106 units/mg. Endotoxin level is less than 0.1 ng per μg of IL-7 (1EU/μg).
  • Protein synonyms:
    IL-7|Interleukin-7
  • Protein/peptide name:
    IL7
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications

About this item

IL-7 is a hematopoietic growth factor, which affects primarily early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors such as ConA and IL-2. Human and murine IL-7 is cross-species reactive. Recombinant human IL-7 is a 17.4 kDa protein containing 153 amino acid residues. Recombinant rat IL-7 is a 15.0 kDa protein containing 130 amino acid residues. Recombinant murine IL-7 is a 15.0 kDa protein containing 130 amino acid residues.

  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.