Order Entry
Sweden
ContactUsLinkComponent
Human 4E-BP1 peptide (14 aa near C-terminus)
Human 4E-BP1 peptide (14 aa near C-terminus)
Catalog # PRSI3513P
Supplier:  ProSci Inc.
CAS Number:  
Human 4E-BP1 peptide (14 aa near C-terminus)
Catalog # PRSI3513P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    14 aa near C-terminus
  • Protein synonyms:
    Eukaryotic Translation Initiation Factor 4EBinding 1|EIF4EBP1
  • Protein/peptide name:
    4E-BP1 Peptide (14 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

4E-BP1 peptide is used for blocking the activity of 4E-BP1 antibody.