Order Entry
Sweden
ContactUsLinkComponent
 
Human TIRP peptide
Catalog # PRSI3206P
Supplier:  ProSci Inc.
CAS Number:  
Human TIRP peptide
Catalog # PRSI3206P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Protein/peptide name:
    TIRP Peptide
  • Tested applications:
    Blocking

Specifications

About this item

TIRP peptide is used for blocking the activity of TIRP antibody.