Anti-CCL7 Goat Polyclonal Antibody
PRSIXP-5223
- Antikroppsnamn:Chemokine (C-C motif) ligand 7
- Antigensymbol:CCL7
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- Reaktivitet:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonymer:MCP3|NC28|SCYA7|SCYA6|MCP-3|MARC|FIC
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant mMCP-3 (murine Macrophage/Monocyte chemotactic protein-3).
- Storlek:100 µg
- Förp.:100 µG
Anti-mMCP-3 specific antibody was purified by affinity chromatography employing immobilized mMCP-3 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CCL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse