Anti-GFP Chicken Polyclonal Antibody
Catalog # PRSIXG-6169
Supplier: ProSci Inc.
Specifications
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Immunogen:GFP fusion protein.
- Size:100 µl
- Pk:500 µG
Specifications
About this item
This antibody can be used at 1:500 to 1:2,000 for both immunoblot and ELISA testing, and 1:200 to 1:1000 for ICC (immunocytochemistry) test. This antibody can be used in combination with Anti-IgY HRP or Anti-IgY AP for ELISA or Western blot analysis and Anti-IgY FITC for ICC.
PBS Buffer containing 0.02% sodium azide.
PBS Buffer containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: gfp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse