Specifications
- Antigen name:Homeobox protein NANOG
- Antigen symbol:NANOG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ENK|ecat4|2410002E02Rik
- Concentration:1 mg/mL
- Immunogen:Nanog antibody was raised against a peptide sequence around aa.137~141 (K-Q-V-K-T) derived from Nano g.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Transcription regulator involved in inner cell mass and embryonic stem (ES) cells proliferation and self-renewal. Imposes pluripotency on ES cells and prevents their differentiation towards extraembryonic endoderm and trophectoderm lineages. Blocks bone morphogenetic protein-induced mesoderm differentiation of ES cells by physically interacting with SMAD1 and interfering with the recruitment of coactivators to the active SMAD transcriptional complexes By similarity. Acts as a transcriptional activator or repressor By similarity. Binds optimally to the DNA consensus sequence 5'-TAAT[GT][GT]-3' or 5'-[CG][GA][CG]C[GC]ATTAN[GC]-3' By similarity. When overexpressed, promotes cells to enter into S phase and proliferation
Western Blot: 1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse