Specifications
- Antigen name:Transient Receptor Potential Cation Channel subfamily V member 1
- Antigen symbol:TRPV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TrpV1|VR1|TRPV1|Transient receptor potential cation channel subfamily V member 1|Capsaicin receptor
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal TRPV1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TRPV1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TRPV1 Antibody: TRPV1 is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. This receptor is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo.
TRPV1 Antibody is affinity chromatography purified via peptide column.
TRPV1 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human