Order Entry
Sweden
ContactUsLinkComponent
Anti-GPR61 Rabbit Polyclonal Antibody
Anti-GPR61 Rabbit Polyclonal Antibody
  PRSI48-302
undefined
Anti-GPR61 Rabbit Polyclonal Antibody
  PRSI48-302

 

  • Antigen name:
    G Protein-coupled Receptor 61
  • Antigen symbol:
    GPR61
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Bat,
    Pig,
    Horse,
    Monkey,
    Gibbon,
    Gorilla,
    Human,
    Bovine,
    Rat,
    Mouse
  • Form:
    Liquid
  • Antigen synonyms:
    BALGR|GPCR3
  • Immunogen:
    GPR61 antibody was raised against a peptide located in the 2nd cytoplasmic domain of GPR61 (Human).
  • Tested applications:
    Immunoaffinity Chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

GPR61 is an Orphan-A GPCR with an unknown ligand.

GPR61 antibody can be used in immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Gorilla, Horse, Human, Monkey, Mouse, Pig, Rat