Anti-APOBEC1 Goat Polyclonal Antibody
PRSI46-761
- Antigen name:Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide 1
- Antigen symbol:APOBEC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:BEDP|CDAR1|APOBEC-1|HEPR
- Concentration:500 ug/mL
- Immunogen:APOBEC1 antibody was raised against a 14 amino acid synthetic peptide near the internal region (near N-Terminus) of APOBEC1.
- Size:100 µg
- Pk:100 µG
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 28 kDa band observed in mouse spleen lysates (calculated MW of 28.2 kDa according to human NP_001635.2 and 27.5 kDa acccording to mouse NP_112436.1). Recommended concentration:1-3 μg/mL.
APOBEC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
APOBEC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOBEC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse