Order Entry
Sweden
ContactUsLinkComponent
Anti-RNF34 Goat Polyclonal Antibody
Anti-RNF34 Goat Polyclonal Antibody
Catalog # PRSI46-283
Supplier:  ProSci Inc.
undefined
Anti-RNF34 Goat Polyclonal Antibody
Catalog # PRSI46-283
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Ring Finger Protein 34
  • Antigen symbol:
    RNF34
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    caspases-8 and -10-associated RING finger protein 1|FLJ21786|RIFF|FYVE-RING finger protein MOMO|RING finger protein RIFF|CARP1|RFI|hRFI|RIF
  • Concentration:
    500 ug/mL
  • Immunogen:
    RFI antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of RFI.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

RFI antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RFI antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RNF34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0