Anti-VHL Goat Polyclonal Antibody
Catalog # PRSI45-170
Supplier: ProSci Inc.
Specifications
- Antigen name:Von Hippel-Lindau Tumor Suppressor
- Antigen symbol:VHL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:VHL|VHLH|RCA1|pVHL|VHL1|HRCA1
- Concentration:500 ug/mL
- Immunogen:VHL antibody was raised against a 13 amino acid synthetic peptide near the internal region of VHL.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
VHL antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
VHL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: VHL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat