Anti-LEF1 Goat Polyclonal Antibody
PRSI45-083
- Antigen name:Lymphoid Enhancer-Binding Factor 1
- Antigen symbol:LEF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TCF1ALPHA|TCF1-alpha|DKFZp586H0919|LEF-1|LEF1
- Concentration:500 ug/mL
- Immunogen:LEF1 antibody was raised against a 14 amino acid synthetic peptide near the internal region of LEF1.
- Size:100 µg
- Pk:100 µG
LEF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LEF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LEF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat