Anti-DND1 Rabbit Polyclonal Antibody
Catalog # PRSI29-529
Leverantör: ProSci Inc.
Specifikationer
- Antigen name:DND MicroRNA-mediated Repression Inhibitor 1
- Antigen symbol:DND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:RBMS4
- Immunogen:A synthetic peptide corresponding a region of human DND1.
- Size:50 µg
- Pk:50 µG
Specifikationer
Om denna produkt
DND1 contains 2 RRM (RNA recognition motif) domains. It may play a role during primordial germ cell (PGC) development. However, DND1 does not seem to be essential for PGC migration
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human