- Antigen name:Minichromosome Maintenance Complex Component 4
- Antigen symbol:MCM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:S. cerevisiae|KIAA4003|DNA replication licensing factor MCM 4|Minichromosome maintenance deficient 4|MCM 4 minichromosome maintenance deficient 4|Minichromosome maintenance|AU045576|mcdc21|P1 CDC21|hCdc21|P1CDC21|MGC33310|Homolog of S. pombe cell devision cycle 21|P1-CDC21|Minichromosome maintenance deficient 4 homolog S. cerevisiae|19G AI325074|4|mitotin S. cerevisiae|mKIAA4003|Minichromosome maintenance deficient S. cerevisiae 4|CDC54|Mcmd4|Minichromosome maintenance complex component 4|MCM4_HUMAN|CDC21|homolog of|CDC 21|MCM4|MCM4 minichromosome maintenance deficient 4|DNA replication licensing factor MCM4|CDC 54.|CDC21 homolog|hCdc 21|Minichromosome Maintenance 4|MCM 4
- Immunogen:A synthetic peptide corresponding a region of human MCM4.
- Size:100 µg
- Pk:100 µG
The protein encoded by MCM4 is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MCM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human