Order Entry
Sweden
ContactUsLinkComponent
Anti-EPB42 Rabbit Polyclonal Antibody
Anti-EPB42 Rabbit Polyclonal Antibody
Catalog # PRSI27-947
Supplier:  ProSci Inc.
undefined
Anti-EPB42 Rabbit Polyclonal Antibody
Catalog # PRSI27-947
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Erythrocyte membrane Protein band 4.2
  • Antigen symbol:
    EPB42
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    PA|SPH5
  • Immunogen:
    A synthetic peptide corresponding a region of human EPB42.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EPB42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat