Anti-LHX3 Rabbit Polyclonal Antibody
Catalog # PRSI27-913
Supplier: ProSci Inc.
Specifications
- Antigen name:LIM Homeobox 3
- Antigen symbol:LHX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:DKFZp762A2013|CPHD3|LIM3|M2-LHX3
- Immunogen:A synthetic peptide corresponding a region of human LHX3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
LHX3 encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LHX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human