Anti-GPBP1 Rabbit Polyclonal Antibody
PRSI27-731
- Antigen name:GC-rich promoter binding protein 1
- Antigen symbol:GPBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:VASCULIN|GPBP|SSH6
- Immunogen:A synthetic peptide corresponding a region of human DKFZP761C169.
- Size:50 µg
- Pk:50 µG
Vasculin is a novel vascular protein differentially expressed in human atherogenesis
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GPBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human