Order Entry
Sweden
ContactUsLinkComponent
Anti-C16ORF78 Rabbit Polyclonal Antibody
Anti-C16ORF78 Rabbit Polyclonal Antibody
Catalog # PRSI26-677
Supplier:  ProSci Inc.
undefined
Anti-C16ORF78 Rabbit Polyclonal Antibody
Catalog # PRSI26-677
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Chromosome 16 open reading frame 78
  • Antigen symbol:
    C16ORF78
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human C16orf78.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The exact function of C16orf78 remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: C16ORF78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human