Order Entry
Sweden
ContactUsLinkComponent
Anti-ZIP5 Rabbit Polyclonal Antibody
Anti-ZIP5 Rabbit Polyclonal Antibody
  PRSI6089
undefined
Anti-ZIP5 Rabbit Polyclonal Antibody
  PRSI6089

 

  • Antigen name:
    Solute carrier Family 39 (metal ion transporter), member 5
  • Antigen symbol:
    ZIP5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    ZIP5 antibody was raised against a 17 amino acid synthetic peptide near the center of human ZIP5.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ZIP5 Antibody: The zinc transporter ZIP5, also known as SLC39A5, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP5 is closely related to ZIP4, and is found in many of the same tissues, including intestine, pancreas, liver, and kidney, but unlike ZIP4, ZIP5 localizes to the basolateral membrane in kidney cells and its activity is not downregulated by zinc repletion. Recent evidence suggests that ZIP4 and ZIP5 are reciprocally regulated, with translation of ZIP5 mRNA decreasing and ZIP5 protein being rapidly internalized and degraded in response to zinc deficiency.

Type:
Antigen: ZIP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse