- Antigen name:Inhibitor of ? light polypeptide gene enhancer in B-cells, kinase complex-associated protein
- Antigen symbol:DYS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:IKI 3|DYS|IKK complex associated protein|ELP1_HUMAN|IKK complex-associated protein|Elongator complex protein 1|IkappaB kinase complex-associated protein|ELP1|ikbkap|DKFZp781H1425|IKI3|Dysautonomia Riley Day syndrome hereditary sensory autonomic neuropathy type III|FD|IKAP|FLJ12497|TOT1|ELP 1|TOT 1|IkappaB kinase complex associated protein
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:IKAP antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human IKAP.
- Tested applications:E, WB, ICC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
IKAP Antibody: IKAP was initially identified as a scaffold protein of the I kappa B kinase complex that could bind to IKK alpha , IKK beta , NF-kappa B, and the NF-kappa B-inducing kinase (NIK), although later evidence has cast doubt on this. More recent reports show that mutations in IKAP such as a frameshift leading to a truncated protein or a missense mutation that leads to defective phosphorylation are responsible for the autosomal recessive genetic disease familial dysautonomia (FD). Reports indicating that it forms part of the RNA polymerase II transcription elongation complex suggest that this disease may be due to compromised transcription elongation. More recently, it was shown that IKAP associates with c-Jun N-terminal kinase (JNK) and could specifically enhance JNK activation induced by the upstream JNK activators MEKK1 and ASK1, indicating another possible cause for FD. At least two isoforms of IKAP are known two exist.
Type:
Antigen: IKAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse