Order Entry
Sweden
ContactUsLinkComponent
Filtration capsules, Sartopore® Platinum
Filtration capsules, Sartopore® Platinum
Supplier:  Sartorius
Certificates
undefined
Filtration capsules, Sartopore® Platinum
Supplier:  Sartorius
Specifications
  • Catalog No:
  • SARP5495307H9-OO-A
  • SARP5495307H8-OO-A
  • Inlet/outlet:
  • 1" Single-stepped hose barb
  • 1" Single-stepped hose barb
  • Ø×L:
  • 77×161 mm
  • 77×161 mm
  • Max. pressure:
  • 5 bar @ 20 °C; 3 @ 50 °C
  • 5 bar @ 20 °C; 3 @ 50 °C
Specifications