Order Entry
Sweden
ContactUsLinkComponent
Chondroitin Sulfate, Reference Standards
Chondroitin Sulfate, Reference Standards
Supplier:  USP
Certificates
Chondroitin Sulfate, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    9082-07-9
  • Description:
    Chondroitin Sulfate Sodium
  • Formula:
    C₁₄H₂₂NNaO₁₆S

Specifications