Anti-IL6 Rabbit Polyclonal Antibody
BSBTA00102-2
- Antibody type:Primary
- Antigen symbol:IL6
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for IL6 detection. Tested with WB, ELISA(Cap) in Mouse;Rat.
Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat