Anti-GRO alpha Rabbit Polyclonal Antibody
BSBTA00533
- Antibody type:Primary
- Antigen name:GRO alpha
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for GRO alpha detection. Tested with WB, ELISA(Cap) in Mouse;Rat.
Type: Primary
Antigen: GRO alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat