Anti-SIX6 Rabbit Polyclonal Antibody
Catalog # BSBTA06502-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Homeobox protein SIX6
- Antigen symbol:SIX6
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Homeobox protein SIX6(SIX6) detection. Tested with WB in Human.
Type: Primary
Antigen: SIX6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human