Anti-MUC2 Rabbit Polyclonal Antibody
BSBTA01212
- Antibody type:Primary
- Antigen symbol:MUC2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for MUC2 detection. Tested with IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: MUC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat