Nitrile gloves, Dexpure® 801-95
Supplier: HONEYWELL SAFETY
Certificates
About this item
These gloves are particularly resistant to animal fat and detergents. They also offer good mechanical resistance, making it an economical alternative to the traditional reusable model. They are ideal for food preparation of greasy products (cooked dishes, pastries, and more.) and cleaning applications in contact with oils, animals and vegetable fat.
- Alternative to latex gloves as they offer a better mechanical resistance than latex reduce the risk of allergic reactions
- Superior performance when in contact with greasy products
- Patterned finish provides a better grip
- Certified for contact with food according to European directive N°1934/2004
- Easily detectable thanks to blue colour
Certifications: EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. II; AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- DALL4580195/7
- DALL4580195/10
- DALL4580195/9
- DALL4580195/8
- Modell:
- 801-95
- 801-95
- 801-95
- 801-95
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured
- Textured
- Textured
- Textured
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- Godkända för livsmedelskontakt:
- +
- +
- +
- +