Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standards, tiotropium compounds, Ph. Eur., RS
Organic reference standards, tiotropium compounds, Ph. Eur., RS
:
EDQM
Organic reference standards, tiotropium compounds, Ph. Eur., RS
:
EDQM
CAS No.:
139404-48-1
Formula:
C₁₉H₂₄BrNO₅S₂
Description:
Tiotropium bromide monohydrate