Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, rosuvastatin, Ph. Eur., RS
Organic reference standards, rosuvastatin, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, rosuvastatin, Ph. Eur., RS
Supplier:  EDQM
Certificates

Specifications

  • CAS No.:
    147098-20-2
  • Formula:
    C₄₄H₅₄CaF₂N₆O₁₂S₂
  • Description:
    Rosuvastatin Calcium

Specifications

Related Information