Impurity and API standard, levamisole hydrochloride
Catalog # PROOMM0706.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:16595-80-5
- Description:Levamisole hydrochloride
- Formula:C₁₁H₁₃ClN₂S
- Description:Levamisole hydrochloride