Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, Quinine sulphate standard, BP
Organic reference standards, Quinine sulphate standard, BP
Catalog # PROOBP862
CAS Number:  
Organic reference standards, Quinine sulphate standard, BP
Catalog # PROOBP862
CAS Number:  

Specifications

  • Pk:
    300 mg
  • Standards:
    Reference standard
  • CAS No.:
    6119-70-6
  • Description:
    Quinine sulphate
  • Formula:
    C₂₀H₂₆N₂O₆S
  • Description:
    Quinine sulphate

Specifications

About this item