Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
VWR®, Accessories for Tester pH/CO 1030
VWR®, Accessories for Tester pH/CO 1030
VWR®, Accessories for Tester pH/CO 1030
Supplier:
VWR Collection
Certificates
Expand image
VWR®, Accessories for Tester pH/CO 1030
Supplier:
VWR Collection
About this item