Organic reference standards, diflucortolone valerate assay standard, BP
Catalog # PROOBP401
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standard
- CAS No.:59198-70-8
- Description:Diflucortolone valerate assay standard
- Formula:C₂₇H₃₆F₂O₅
- Description:Diflucortolone valerate assay standard