Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, ofloxacin, Ph. Eur. RS
Organic reference standards, ofloxacin, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, ofloxacin, Ph. Eur. RS
Supplier:  EDQM
Certificates

Specifications

  • CAS No.:
    82419-36-1
  • Formula:
    C₁₈H₂₀FN₃O₄
  • Description:
    Ofloxacin

Specifications

Related Information