Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, isradipines, Ph. Eur.
Organic reference standards, isradipines, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, isradipines, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    75695-93-1
  • Formula:
    C₁₉H₂₁N₃O₅
  • Description:
    Isradipine

Specifications