Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, gemcitabines, Ph. Eur.
Organic reference standards, gemcitabines, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, gemcitabines, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    122111-03-9
  • Formula:
    C₉H₁₂ClF₂N₃O₄
  • Description:
    Gemcitabine hydrochloride

Specifications