Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, furosemides, Ph. Eur.
Organic reference standards, furosemides, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, furosemides, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    54-31-9
  • Formula:
    C₁₂H₁₀ClN₂O₅S
  • Description:
    Furosemide

Specifications