
Natriumhydroxid 1.0 N i vattenlösning, Biotech Reagent, J.T.Baker®
: Avantor
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:1,04 g/cm³ (25 °C)
:Omgivande
:215-185-5
:3266
:8,III
:Filtered through a 0.2 µm filter.
:GHS05
- GMP Manufactured Product:0.975 - 1.025
- CAUTION: For Manufacturing, processing or repackaging:≤5 ppm
- Appearance (clear, colorless solution):Passes Test
- Normality:0.975 - 1.025
- Chloride (Cl):≤ 5 ppm
- ACS - Heavy Metals (as Pb):≤ 1 ppm
- Trace Impurities - Iron (Fe):≤ 0.5 ppm
- Endotoxin Concentration (EU/mL):≤ 2.0