Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Frätande
Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Leverantör:  Avantor
Certifikat
undefined
Frätande
Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Leverantör:  Avantor
Certifikat

Specifikationstestresultat

  • GMP Manufactured Product:
    0.480 - 0.520
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤5 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1 ppm
  • Appearance (clear, colorless solution):
    Passes Test
  • Normality:
    0.480 - 0.520
  • Chloride (Cl):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.5 ppm
  • Endotoxin Concentration (EU/mL):
    ≤ 2.0

Specifikationstestresultat

Relaterad information

Certifikat
Sök efter LOT-nummer