Order Entry
Sweden
ContactUsLinkComponent
Sodium dihydrogen phosphate dihydrate Ph. Eur., Macron Fine Chemicals™
Sodium dihydrogen phosphate dihydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P108-42
CAS Number:  
Sodium dihydrogen phosphate dihydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P108-42
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

About this item

 :
 :156.01 g/mol
CAS Number:13472-35-0
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209