Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propylene glycol
(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™
(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™
(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
Expand image
(±)-1,2-Propanediol Ph. Eur., Macron Fine Chemicals™
Supplier:
MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:
57-55-6
:
Formula:
CH₃CH(OH)CH₂OH
MDL Number:
MFCD00064272
:
76.1 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
BAKRP148-38_TDS_EN
SDS
sds_MACRP148
Documentation