

Orthophosphoric acid ≥85.0-88.0% (by acidimetry) FCC, NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Bulk Food Chemical:85.0 - 88.0 %
- NF - Assay (H₃PO₄) (by acidimetry):85.0 - 88.0 %
- NF - Alkali Phosphates:Passes Test
- NF - Identification:Passes Test
- NF - Nitrate (NO₃):Passes Test
- NF - Phosphorous or Hypophosphorous Acid:Passes Test
- NF - Sulfate (SO₄):Passes Test
- FCC - Assay (H₃PO₄) (by acidimetry):85.0 - 88.0 %
- FCC - Arsenic (As):≤3 mg/kg
- FCC - Cadmium (Cd):≤3 mg/kg
- FCC - Fluoride (F):≤10 mg/kg
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤3 mg/kg