Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid ≥85.0-88.0% NF, FCC, J.T.Baker®
Corrosive
Irritant
Orthophosphoric acid ≥85.0-88.0% NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Orthophosphoric acid ≥85.0-88.0% NF, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Food GMP Manufactured Product:
    85.0 - 88.0%
  • GMP Manufactured Product:
    Passes Test
  • Meets N.F. Requirements:
    Passes Test
  • Meets F.C.C. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    85.0 - 88.0%
  • NF - Assay (H₃PO₄):
    85.0 - 88.0 %
  • NF - Identification:
    Passes Test
  • NF - Nitrate (NO₃):
    Passes Test
  • NF - Phosphorous or Hypophosphorous Acid:
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Alkali Phosphates:
    Passes Test
  • FCC - Assay (H₃PO₄):
    85.0 - 88.0 %
  • FCC - Identification:
    Passes Test
  • FCC - Arsenic (As):
    ≤3 mg/kg
  • FCC - Cadmium (Cd):
    ≤3 mg/kg
  • FCC - Lead (Pb):
    ≤3 mg/kg
  • FCC - Fluoride (F):
    ≤10 mg/kg

Specification Test Results

Related Information