Order Entry
Sweden
ContactUsLinkComponent
2-Propanol ≥99.0% (by GC) USP, Macron Fine Chemicals™
2-Propanol ≥99.0% (by GC) USP, Macron Fine Chemicals™
Leverantör:  MACRON AVANTOR BRAND
Certifikat
undefined
2-Propanol ≥99.0% (by GC) USP, Macron Fine Chemicals™
Leverantör:  MACRON AVANTOR BRAND
Certifikat

Specifikationstestresultat

  • Bulk Pharmaceutical Chemical:
    = 99.0 %
  • USP - Assay (CH₃CHOHCH₃) (by GC):
    ≥99.0 %
  • USP - Appearance (clear, colorless liquid):
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Specific Gravity at 25°/25°C:
    0.783 - 0.787
  • USP - Acidity (mL):
    ≤0.70
  • USP - Nonvolatile Residue:
    ≤0.005 %
  • USP - Refractive Index, n²ºD:
    1.376 - 1.378
  • USP - Water (H₂O):
    ≤0.5 %
  • USP - Volatile Impurities - Ethyl ether:
    ≤0.1 %
  • USP - Volatile Impurities - Acetone:
    ≤0.1 %
  • USP - Volatile Impurities - Diisopropyl ether:
    ≤0.1 %
  • USP - Volatile Impurities - 1-Propanol:
    ≤0.1 %
  • USP - Volatile Impurities - 2-Butanol:
    ≤0.1 %
  • USP - Volatile Impurities - Total:
    ≤1.0 %
  • USP - Benzene:
    ≤2 ppm

Specifikationstestresultat

Relaterad information