Order Entry
Sweden
ContactUsLinkComponent
Sodium hypochlorite 0.5 - 0.7% available chlorine, J.T.Baker®
Sodium hypochlorite 0.5 - 0.7% available chlorine, J.T.Baker®
Catalog # 3917.
Supplier:  Avantor
CAS Number:  
Sodium hypochlorite 0.5 - 0.7% available chlorine, J.T.Baker®
Catalog # 3917.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Active Chlorine:
    0.5 - 0.7 %

Specification Test Results

About this item

 :
 :74.44 g/mol
UN:3212
ADR:5.1,II
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120