
Guanidinium hydrochloride 99.5-101.0%, NF-GenAR® for biotechnology, Macron Fine Chemicals™
Catalog # 0506-06
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE pail, screw top
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- GMP Manufactured Product:≤0.3%
- CAUTION: For Manufacturing, processing or repackaging:99.5 - 101.0%
- Appearance:Passes Test
- Water-Insoluble Matter:≤0.3 %
- Assay (NH₂C(:NH)NH₂·HCl):99.5 - 101.0 %
- Loss on Drying:≤0.5 %
- Trace Impurities - Iron (Fe):≤2.0 ppm
Specification Test Results
About this item
:
:95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026