Glycerine Ph. Eur., Macron Fine Chemicals™
Catalog # P137-33
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:92.09 g/mol
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722