Order Entry
Sweden
ContactUsLinkComponent
Aluminium potassium sulphate dodecahydrate Ph. Eur., Macron Fine Chemicals™
Aluminium potassium sulphate dodecahydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P124-38
Supplier:  Avantor
CAS Number:  
Aluminium potassium sulphate dodecahydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P124-38
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :474.39 g/mol
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143