Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, arbutin, Ph. Eur.
Organic reference standards, arbutin, Ph. Eur.
Katalog # EDQMY0000806
Leverantör:  EDQM
CAS-nummer:  
Organic reference standards, arbutin, Ph. Eur.
Katalog # EDQMY0000806
Leverantör:  EDQM
CAS-nummer:  

Specifikationer

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    497-76-7
  • Description:
    Arbutin, Ph. Eur. standard
  • Formula:
    C₁₂H₁₆O₇
  • Description:
    Arbutin

Specifikationer

Om denna produkt